missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA2G4/EBP1 Antibody (5E7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59468JF549
This item is not returnable.
View return policy
Description
PA2G4/EBP1 Monoclonal antibody specifically detects PA2G4/EBP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| PA2G4/EBP1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Cell cycle protein p38-2G4 homolog, EBP1, ErbB-3 binding protein 1, ErbB3-binding protein 1, ErbB3-binding protein Ebp1, hG4-1, p38-2G4, proliferation-associated 2G4, 38kD, proliferation-associated 2G4, 38kDa, proliferation-associated protein 2G4 | |
| Recombinant human PA2G4/EBP1 (1-394aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 5036 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 50000000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction