missing translation for 'onlineSavingsMsg'
Learn More

PA28 Activator gamma Subunit/PSME3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18373632 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373632 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373632 Supplier Novus Biologicals Supplier No. NBP300356JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA28 Activator gamma Subunit/PSME3 Polyclonal specifically detects PA28 Activator gamma Subunit/PSME3 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PA28 Activator gamma Subunit/PSME3
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias 11S regulator complex gamma subunit, 11S regulator complex subunit gamma, Activator of multicatalytic protease subunit 3, Ki, Ki antigen, PA28g, PA28gamma, PA28-gamma, PA28GKi nuclear autoantigen, proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki), Proteasome activator 28 subunit gamma, proteasome activator 28-gamma, proteasome activator complex subunit 3, REG-gamma
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PA28 Activator gamma Subunit/PSME3 (P61289-1; Met1-Tyr254).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Immune System Diseases, Immunology, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 10197
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.