missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA28 Activator beta Subunit/PSME2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£155.00
Specifications
| Antigen | PA28 Activator beta Subunit/PSME2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18526542
|
Novus Biologicals
NB120-2940-0.025ml |
0.025 mL | |||||||
Description
PA28 Activator beta Subunit/PSME2 Polyclonal specifically detects PA28 Activator beta Subunit/PSME2 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| PA28 Activator beta Subunit/PSME2 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10 - 1:100 | |
| Polyclonal | |
| Precipitation | |
| RUO | |
| Human | |
| 11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta | |
| PSME2 | |
| IgG | |
| Ammonium sulfate precipitation | |
| Detects recombinant human proteasome 11S REG beta. This does not detect endogenous levels of proteasome 11S REG beta nor does it detect other 11S REG subunits. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS with 0.05% Sodium Azide | |
| 5721 | |
| Synthetic Peptide: M(1) A K P C G V R L S G E A R(14) | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title