missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA28 Activator beta Subunit/PSME2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35372-20ul
This item is not returnable.
View return policy
Description
PA28 Activator beta Subunit/PSME2 Polyclonal antibody specifically detects PA28 Activator beta Subunit/PSME2 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| PA28 Activator beta Subunit/PSME2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| 11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-239 of human PA28 Activator beta Subunit/PSME2 (NP_002809.2).,, Sequence:, MAKPCGVRLSGEARKQVEVFRQNLFQEAEEFLYRFLPQKIIYLNQLLQEDSLNVADLTSLRAPLDIPIPDPPPKDDEMETDKQEKKEVHKCGFLPGNEKVLSLLALVKPEVWTLKEKCILVITWIQHLIPKIEDGNDFGVAIQEKVLERVNAVKTKVEAFQTTISKYFSERGDAVAKASKETHVMDYRALVHERDEAAYGELRAMVLDLRAFYAELYHIISSNLEKIVNPKGEEKPSMY | |
| 20 μL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5721 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction