missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA28 Activator beta Subunit/PSME2 Antibody (06), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06553-50ul
This item is not returnable.
View return policy
Description
PA28 Activator beta Subunit/PSME2 Monoclonal antibody specifically detects PA28 Activator beta Subunit/PSME2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| PA28 Activator beta Subunit/PSME2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| 06 | |
| Western Blot 1:500-1:2000, ELISA 1:1000-1:2000, Immunoprecipitation 1-4 uL/mg of lysate | |
| 11S regulator complex beta subunit, 11S regulator complex subunit beta, Activator of multicatalytic protease subunit 2, MCP activator, 31-kD subunit, PA28b, PA28betacell migration-inducing protein 22, proteasome (prosome, macropain) activator subunit 2 (PA28 beta), Proteasome activator 28 subunit beta, proteasome activator 28-beta, proteasome activator complex subunit 2, proteasome activator hPA28 subunit beta, REGbeta, REG-beta | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PA28 Activator beta Subunit/PSME2 (Accession#: Met1-Tyr239). | |
| 50 μg | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5721 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction