missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PA28 Activator alpha Subunit/PSME1 Antibody (016), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90298C
This item is not returnable.
View return policy
Description
PA28 Activator alpha Subunit/PSME1 Monoclonal specifically detects PA28 Activator alpha Subunit/PSME1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| PA28 Activator alpha Subunit/PSME1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 5720 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 16 | |
| Western Blot, ELISA | |
| 11S regulator complex alpha subunit, 11S regulator complex subunit alpha, 29-kD MCP activator subunit, IFI5111MGC8628, IGUP I-5111, Interferon gamma up-regulated I-5111 protein, interferon-gamma IEF SSP 5111, interferon-gamma-inducible protein 5111, PA28a, PA28alphaActivator of multicatalytic protease subunit 1, proteasome (prosome, macropain) activator subunit 1 (PA28 alpha), Proteasome activator 28 subunit alpha, proteasome activator complex subunit 1, proteasome activator subunit-1, REGalpha, REG-alpha | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human PA28 Activator alpha Subunit/PSME1 (Q06323; Met1-Tyr249). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction