missing translation for 'onlineSavingsMsg'
Learn More

PA1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18416433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416433 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416433 Supplier Novus Biologicals Supplier No. NBP176239FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1 Polyclonal antibody specifically detects PA1 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen PA1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias chromosome 16 open reading frame 53, FLJ22459, GAS, glutamate-rich coactivator interacting with SRC1/NCOA1, MGC4606, PA1, PAXIP1-associated protein 1, PTIP-associated 1 protein, PTIP-associated protein 1
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human GAS. The immunogen is located within the last 50 amino acids of GAS. Amino Acid Squence: LDSEDPSPASPPLRS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 79447
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.