missing translation for 'onlineSavingsMsg'
Learn More

PAβN dihydrochloride, MedChemExpress

Product Code. 16479845
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479845 250 mg 250mg
16493368 10 mg 10mg
16489896 50 mg 50mg
16462736 25 mg 25mg
16436097 5 mg 5mg
16414616 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16479845 Supplier Medchem Express Supplier No. HY101444A250mg
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.

PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.

Specifications

Chemical Name or Material PAβN dihydrochloride
Physical Form Solid
CAS 100929-99-5
Color White
Quantity 250 mg
Purity Grade Notes Research
Molecular Formula C25H32Cl2N6O2
For Use With (Application) COVID-19-immunoregulation
Synonym MC-207,110 dihydrochloride Phe-Arg-β-naphthylamide dihydrochloride
Solubility Information DMSO : 83.33 mg/mL (160.41 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (27.51 mM; Need ultrasonic)
SMILES O=C([C@H](CCCNC(N)=N)NC([C@H](CC1=CC=CC=C1)N)=O)NC2=CC(C=CC=C3)=C3C=C2.Cl.Cl
Molecular Weight (g/mol) 519.47
Formula Weight 519.47
Percent Purity 98.51%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.