missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAβN dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
25mg
50mg
5mg
Description
PAβN dihydrochloride (MC-207110 dihydrochloride) is an efflux pump inhibitor.
Specifications
Specifications
| Chemical Name or Material | PAβN dihydrochloride |
| Physical Form | Solid |
| CAS | 100929-99-5 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C25H32Cl2N6O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | MC-207,110 dihydrochloride Phe-Arg-β-naphthylamide dihydrochloride |
| Solubility Information | DMSO : 83.33 mg/mL (160.41 mM; Need ultrasonic) ∣H2O : 14.29 mg/mL (27.51 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction