missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PA Janelia Fluor™ 646, Haloalkane
Click to view available options
:
10 μg
Description
Key Information: PA Janelia Fluor™ 646, Haloalkane is a red cell-permeable photoactivable fluorescent dye with a chloroalkane handle for labeling fusion tag proteins.
Application: Live-cell imaging as a self-labeling tag substrate. Suitable for confocal microscopy, super-resolution microscopy (SRM) techniques including PALM (in both live and fixed cells). Allows multicolor single tracking experiments.
Properties and Photophysical Data: PA Janelia Fluor™ 646, Haloalkane shows superior brightness and photostability.
Janelia Fluor is a registered trademark of Howard Hughes Medical Institute.
Specifications
Specifications
| Chemical Name or Material | 2',7'-Bis(1-azetidinyl)-2-diazo-N-[2-[2-[(6-chlorohexyl)oxy]ethoxy]ethyl]-2,3-dihydro-9',9'-dimethyl-3-oxospiro[1H-indene-1,10'(9'H)-[9]silaanthracene]-6-carboxamide |
| CAS | 2011720-21-9 |
| Quantity | 10 μg |
| Target | Fluorescent Dyes |
| Molecular Formula | C40H48ClN5O4Si |
| Purity | 0.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction