missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PA 8

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PA 8 is a PAC1 receptor antagonist. Inhibits PACAP-induced CREB phosphorylation in PAC1, but not VPAC1 or VPAC2, expressing cells. Also inhibits PACAP-induced increases in cAMP levels in vitro (IC50 = 2 nM) and PACAP-induced aversive responses and mechanical allodynia following intrathecal injection in vivo.
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5,8-dihydro-5-[3-methoxy-4-(2-propen-1-yloxy)phenyl]pyrido[2,3-d]pyrimidine-4,7(3H,6H)-dione |
| CAS | 878437-15-1 |
| Quantity | 10 mg |
| Molecular Formula | C17H18N4O4 |
| Formula Weight | 342.35 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction