missing translation for 'onlineSavingsMsg'
Learn More

p97/VCP Rabbit anti-Human, Mouse, Rat, Clone: 3P8T3, Novus Biologicals™

Product Code. 18374352
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374352 20 μg 20µL
18331627 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18374352 Supplier Bio-Techne Supplier No. NBP31547820UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p97/VCP Monoclonal antibody specifically detects p97/VCP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen p97/VCP
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 3P8T3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias 15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human p97/VCP (P55072).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cellular Markers, DNA Repair, Membrane Trafficking and Chaperones, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7415
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.