missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p97/VCP Antibody (5H12), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | p97/VCP |
|---|---|
| Clone | 5H12 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30027687
|
Novus Biologicals
NBP3-26257-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30035238
|
Novus Biologicals
NBP3-26257-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p97/VCP Monoclonal antibody specifically detects p97/VCP in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, ImmunocytochemistrySpecifications
| p97/VCP | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| 15S Mg(2+)-ATPase p97 subunit, IBMPFDMGC8560, MGC131997, p97, TER ATPase, TERA, transitional endoplasmic reticulum ATPase, valosin containing protein, Valosin-containing proteinMGC148092, yeast Cdc48p homolog | |
| A synthesized peptide derived from Human p97/VCP [UniProt P55072] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5H12 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, DNA Repair, Membrane Trafficking and Chaperones, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7415 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title