missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p8 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA14177

Product Code. 11528750

  • £438.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA1-4177 detects human p8 (com1). PA1-4177 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~9 kDa protein representing Human p8 (com1) in HepGe cell lysates. The PA1-4177 immunogen is a synthetic peptide corresponding to residues Q(13) P P G P E D E D S S L D E S D(28) of human p8.

The human p8 gene encodes a nuclear protein with strong mitogenic activity, which is expressed in the pancreas in response to acute pancreatitis. Response to TNF alpha and staurosporine-induced apoptosis is not modified by p8 expression. However, cells expressing p8 grow significantly and more rapidly.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

p8
Polyclonal
Unconjugated
NUPR1
2310032H04Rik; 60B8AG; CAGA; Candidate of metastasis 1; CFAG; CGLA; COM1; CP-10; L1Ag; MA387; MIF; MRP8; NIF; Nuclear protein 1; nuclear protein 1, transcriptional regulator; nuclear protein 1-like; nuclear protein transcription regulator 1; nuclear protein, transcriptional regulator, 1; nuclear proten 1; nuclear transcriptional regulator protein 1; NUPR1; P8; protein p8
Rabbit
Antigen affinity chromatography
RUO
26471
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
O60356
NUPR1
Synthetic peptide corresponding to residues Q(13) P P G P E D E D S S L D E S D(28) of human p8.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.