missing translation for 'onlineSavingsMsg'
Learn More

P7C3, MedChemExpress

Product Code. 16436948
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436948

Brand: Medchem Express HY15976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress P7C3 is an orally bioavailable and blood-brain barrier penetrant aminopropyl carbazole, with neuroprotective effects. P7C3 can be used for the research of neurodegenerative diseases, including Parkinson's disease.

P7C3 is an orally bioavailable and blood-brain barrier penetrant aminopropyl carbazole, with neuroprotective effects. P7C3 can be used for the research of neurodegenerative diseases, including Parkinson's disease.

Specifications

Chemical Name or Material P7C3
CAS 301353-96-8
Color White
Purity Grade Notes Research
Molecular Formula C21H18Br2N2O
Quantity 10 mg
Solubility Information DMSO : ≥ 33 mg/mL (69.59 mM)
SMILES OC(CNC1=CC=CC=C1)CN2C3=CC=C(Br)C=C3C4=CC(Br)=CC=C24
Molecular Weight (g/mol) 474.19
Formula Weight 474.19
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.