missing translation for 'onlineSavingsMsg'
Learn More

P7C3, MedChemExpress

Product Code. 16499487
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499487 5 mg 5mg
16436948 10 mg 10mg
16427026 100 mg 100mg
16413007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499487 Supplier Medchem Express Supplier No. HY15976
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress P7C3 is an orally bioavailable and blood-brain barrier penetrant aminopropyl carbazole, with neuroprotective effects. P7C3 can be used for the research of neurodegenerative diseases, including Parkinson's disease.

P7C3 is an orally bioavailable and blood-brain barrier penetrant aminopropyl carbazole, with neuroprotective effects. P7C3 can be used for the research of neurodegenerative diseases, including Parkinson's disease.

Specifications

Chemical Name or Material P7C3
CAS 301353-96-8
Color White
Purity Grade Notes Research
Molecular Formula C21H18Br2N2O
Quantity 5 mg
Solubility Information DMSO : ≥ 33 mg/mL (69.59 mM)
SMILES OC(CNC1=CC=CC=C1)CN2C3=CC=C(Br)C=C3C4=CC(Br)=CC=C24
Molecular Weight (g/mol) 474.19
Formula Weight 474.19
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.