missing translation for 'onlineSavingsMsg'
Learn More

P7C3-A20, MedChemExpress

Product Code. 16446948
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446948 10 mg 10mg
16423516 25 mg 25mg
16423007 50 mg 50mg
16409497 5 mg 5mg
16437026 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16446948 Supplier Medchem Express Supplier No. HY15978
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress P7C3-A20 is a derivative of P7C3 with potent proneurogenic and neuroprotective activity. P7C3-A20 exerts an antidepressant-like effect. P7C3-A20 can cross the blood-brain barrier and therefore has the potential for brain injury treatment.

P7C3-A20 is a derivative of P7C3 with potent proneurogenic and neuroprotective activity. P7C3-A20 exerts an antidepressant-like effect. P7C3-A20 can cross the blood-brain barrier and therefore has the potential for brain injury treatment.

Specifications

Chemical Name or Material P7C3-A20
CAS 1235481-90-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H19Br2FN2O
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (197.55 mM)
SMILES FC(CNC1=CC(OC)=CC=C1)CN2C3=CC=C(Br)C=C3C4=CC(Br)=CC=C24
Molecular Weight (g/mol) 506.21
Formula Weight 506.21
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.