missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p73 Recombinant Rabbit Monoclonal Antibody (24GB2040)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557809
This item is not returnable.
View return policy
Description
p73 Recombinant Monoclonal Antibody for Western Blot, Flow
P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
Specifications
| p73 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TP73 | |
| deltaNp73; OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; P73; RP23-254N4.10; TAp73; Tp73; transformation related protein 73; Trp73; Tumor protein p73 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 7161 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2040 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15350 | |
| TP73 | |
| A synthesized peptide derived from human p73 (aa 55-80). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction