missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p73 Antibody (ER-15), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB600-1279
This item is not returnable.
View return policy
Description
p73 Monoclonal specifically detects p73 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Gel Super Shift Assays.
Specifications
| p73 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Gel Super Shift Assays 1:1 - 1:100 | |
| p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Gel Shift | |
| ER-15 | |
| Unconjugated | |
| O15350 | |
| TP73 | |
| Purified monkey recombinant p73alpha protein (aa380-637) | |
| 500 μL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 7161 | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction