missing translation for 'onlineSavingsMsg'
Learn More

p73 Antibody (5B429), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18429856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429856 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429856 Supplier Novus Biologicals Supplier No. NBP224737MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p73 Monoclonal antibody specifically detects p73 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen p73
Applications Flow Cytometry
Classification Monoclonal
Clone 5B429
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Flow (Intracellular)
Formulation 50mM Sodium Borate
Gene Alias p53-related protein, P73p53-like transcription factor, TA p73, TAp73, tumor protein p73
Host Species Mouse
Immunogen This antibody was raised against full-length human p73. It reacts with an epitope that is located on the N-terminal region of human p73. It does not cross react with p53.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7161
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.