missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70(S6K) Mouse anti-Human, Mouse, Rat, Clone: 1D11C6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66638-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the RSK (ribosomal S6 kinase) family of serine/threonine kinases. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates several residues of the S6 ribosomal protein. The kinase activity of this protein leads to an increase in protein synthesis and cell proliferation. Amplification of the region of DNA encoding this gene and overexpression of this kinase are seen in some breast cancer cell lines. Alternate translational start sites have been described and alternate transcriptional splice variants have been observed but have not been thoroughly characterized.Specifications
| p70(S6K) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23443, P67999, Q8BSK8 | |
| RPS6KB1 | |
| p70(S6K) Fusion Protein Ag5883 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1D11C6 | |
| Unconjugated | |
| RPS6KB1 | |
| p70 alpha, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6K, p70 S6K 1, p70 S6K alpha, p70 S6KA, p70(S6K), p70(S6K) alpha, p70S6K, P70S6K1, PS6K, Ribosomal protein S6 kinase I, RPS6KB1, S6K, S6K beta 1, S6K1, STK14A | |
| Mouse | |
| Protein G | |
| RUO | |
| 6198, 72508, 83840 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction