missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70 S6 Kinase/S6K, p Ser411 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£194.00 - £316.00
Specifications
| Antigen | p70 S6 Kinase/S6K (p Ser411) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246341
|
Novus Biologicals
NB100-82120-0.025ML |
0.025 mL |
£194.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18107249
|
Novus Biologicals
NB100-82120 |
0.05 mL |
£316.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| p70 S6 Kinase/S6K (p Ser411) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Autophagy, mTOR Pathway, Phospho Specific, Protein Kinase | |
| P23443 | |
| 6198 | |
| IgG | |
| Affinity Purified | |
| 80 kDa |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Mouse, Human, Rat | |
| EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A | |
| RPS6KB1 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit