Learn More
p70 S6 Kinase/S6K Antibody (215247) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB8962Z
440.30 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18554618
  • £542.00 / 0.10mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
p70 S6 Kinase/S6K Monoclonal antibody specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p70 S6 Kinase/S6K
Monoclonal
DyLight 755
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF
215247
Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A
E. coli-derived recombinant human p70 S6 Kinase, Accession # M60725
0.1 mL
Autophagy, Cancer, mTOR Pathway, Protein Kinase, Signal Transduction
6198
Store at 4°C in the dark.
IgG1