missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX p70 S6 Kinase Protein, Active

Product Code. 17380234
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17380234

Brand: Eurofins DiscoverX 14486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-terminal His6-tagged, recombinant amino acids 1-421 of human p70 S6 kinase, for use in Kinase Assays.

p70 S6 Kinase, also kown as RPS6KB1 (Ribosomal Protein S6 Kinase B1) is a Protein Coding gene. This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer.

GenBank M60724. The recombinant protein contains an amino acid substitution, T412E, with reference to M60724. This mutation mimics phosphorylation of T412 and facilitates activation by PDK1.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 270 mM sucrose, 150 mM NaCl, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution.
Molecular Weight (g/mol) 48.7 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥67%
Protein p70 S6 Kinase (1-421, T412E), active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.