missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p70 S6 Kinase beta/S6K2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | p70 S6 Kinase beta/S6K2 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18453792
|
Novus Biologicals
NBP1-87805-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18204677
|
Novus Biologicals
NBP1-87805 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
p70 S6 Kinase beta/S6K2 Polyclonal specifically detects p70 S6 Kinase beta/S6K2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| p70 S6 Kinase beta/S6K2 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6199 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VDSPDDTALSESANQAFLGFTYVAPSVLDSIKEGFSFQPKLRSPRRLNSSPRAPVSPLKFSPFEGFRPSPSLPEPTELPLPPLLPPPPPSTTAPLPIRPPSGTKK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14BS6 kinase-related kinase | |
| RPS6KB2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title