missing translation for 'onlineSavingsMsg'
Learn More

p70 S6 Kinase beta/S6K2 Antibody (4B11), Novus Biologicals™

Product Code. 18341039 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18341039 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18341039 Supplier Novus Biologicals Supplier No. H00006199M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

p70 S6 Kinase beta/S6K2 Monoclonal antibody specifically detects p70 S6 Kinase beta/S6K2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen p70 S6 Kinase beta/S6K2
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4B11
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH06106
Gene Alias EC 2.7.11, EC 2.7.11.1, KLS, p70 ribosomal S6 kinase beta, p70 S6 kinase beta, p70 S6KB, p70 S6K-beta, p70(S6K)-beta, p70-beta, P70-beta-1, P70-beta-2, p70-S6K 2, P70S6K2, p70S6Kb, ribosomal protein S6 kinase beta-2, ribosomal protein S6 kinase, 70kD, polypeptide 2,70 kDa ribosomal protein S6 kinase 2, ribosomal protein S6 kinase, 70kDa, polypeptide 2, S6K2, S6K-beta, S6K-beta2, S6K-beta-2, serine/threonine-protein kinase 14 beta, Serine/threonine-protein kinase 14B, SRK, STK14BS6 kinase-related kinase
Host Species Mouse
Immunogen RPS6KB2 (AAH06106, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAVFDLDLETEEGSEGEGEPELSPADACPLAELRAAGLEPVGHYEEVELTETSVNVGPERIGPHSFELLRVLGKGGYGKVFQVRKVQGTNLGKIYAMKV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6199
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.