missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
p67phox/NOXA2 Monoclonal antibody specifically detects p67phox/NOXA2 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | p67phox/NOXA2 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 7J7N3 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | autosomal 2, NADPH oxidase activator 2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2FLJ93058, p67phox, p67-phox |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human p67phox/NOXA2 (P19878). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?