missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p67phox/NOXA2 Antibody (001), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90316IR
This item is not returnable.
View return policy
Description
p67phox/NOXA2 Monoclonal specifically detects p67phox/NOXA2 in Human samples. It is validated for ELISA.
Specifications
| p67phox/NOXA2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 4688 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 1 | |
| ELISA | |
| autosomal 2, NADPH oxidase activator 2, NCF-2, neutrophil cytosol factor 2, neutrophil cytosolic factor 2, neutrophil cytosolic factor 2 (65kD, chronic granulomatous disease, autosomal2), NOXA2FLJ93058, p67phox, p67-phox | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human p67phox/NOXA2 (AAH01606.1; Met1-Val526). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction