missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p65/RELA Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | p65/RELA |
|---|---|
| Concentration | 0.52 mg/mL |
| Applications | Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817333
|
Proteintech
10745-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807333
|
Proteintech
10745-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the N terminus of human RELA.
The transcription factor NFkB is widely recognized as a critical mediator of immune and inflammatory responses. In most cell types, NFkB is found in the cytoplasm where it is associated with an inhibitory protein known as IkB. An impressive variety of stimuli (tumor necrosis factor, interleukin 1, T-cell activation signals, bacterial endotoxins, viral transforming proteins, certain growth factors and reactive oxygen intermediates) lead to the rapid nuclear assumulation of NFkB by the induced phosphorylation and subsquent degradation of IkB. In the nucleus, NFkB regulates genes encoding cytokines, cytokine receptors, cell adhesion molecules, proteins involved in coagulation and genes involved in cell growth control. Additionally, NFkB is thought to be an important transcriptional regulator for HIV. Growing evidence indicates that the dysregulation of NFkB may be key to a number of diseases including arthritis and other inflammatory diseases, Alzheimer′s disease, atherosclerosis, and cancer.Specifications
| p65/RELA | |
| Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Pig, Rat, Mouse | |
| Q04206, Q04207 | |
| 100135665, 19697, 309165, 5970 | |
| p65; RELA Fusion Protein Ag1199 | |
| Primary | |
| -20°C | |
| RELA |
| 0.52 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| NFKB 3, NFKB p65, NFKB3, p65, RELA, Transcription factor p65 | |
| RELA | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title