missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p63 (Squamous, Basal & Myoepithelial Cell Marker) Mouse, Clone: TP63/2427, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £423.29
Specifications
| Antigen | p63 (Squamous, Basal & Myoepithelial Cell Marker) |
|---|---|
| Clone | TP63/2427 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
Specifications
| p63 (Squamous, Basal & Myoepithelial Cell Marker) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human p63 protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 63kDa |
| TP63/2427 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 8626 | |
| IgG2b κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Nuclear | |
| HEK293 cells or Prostate Carcinoma or Lung or bladder squamous cell carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title