Learn More
p63 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12143-1-AP-20UL
Description
The p63 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human p63. This antibody recognizes human, mouse, rat antigen. The p63 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
| p63 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 8626 | |
| IgG |
| ChIP Assay, ELISA, Immunofluorescence, Immunoprecipitation | |
| 240 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat, Mouse |