Learn More
p63 (deltaN) Rabbit, anti-Human, Unconjugated, Clone: Poly6190, STEMCELL Technologies
Rabbit Monoclonal Antibody
Brand: Stemcell Technologies 60154
This item is not returnable.
View return policy
Description
- The Poly6190 antibody reacts with the deltaN isoforms of human p63, a member of the p53 family of transcription factors
- Alternative splicing of the p63 transcript gives rise to two classes of isoforms
- The TA isoforms (TA p63-α, -β, -γ) possess an N-terminal transactivation domain, while the deltaN isoforms (deltaN p63-α, -β, -γ) lack the transactivation domain
- The TA isoforms are capable of transactivating p53 targets, whereas they antagonize p53
- The deltaN isoforms are generally dominant to the TA isoforms and repress the latter
- The p63 protein is localized to the nucleus and plays an important role in epithelial stem cell regeneration, epidermal morphogenesis, limb development, and stratification
Specifications
| p63 (deltaN) | |
| Monoclonal | |
| Unconjugated | |
| 200 μL | |
| Human | |
| Airway Cells, Dermal Cells |
| Immunohistochemistry, Western Blot, Immunocytochemistry, Immunofluorescence | |
| Poly6190 | |
| Rabbit | |
| Primary | |
| IgG |