missing translation for 'onlineSavingsMsg'
Learn More

P62/SQSTM1 Mouse anti-Human, Clone: 1H5C1, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen P62/SQSTM1
Clone 1H5C1
Concentration 1.33 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16887825
View Documents
Proteintech
66184-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16877825
View Documents
Proteintech
66184-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The molecular weight of p62 is predicted as 48/ 38 kDa, while western blot analyses using this antibody demonstrate the major band around 60-62 kDa in various tissues.

This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
Specifications

Specifications

P62/SQSTM1
1.33 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A170, EBIAP, ORCA, OSIL, p60, P62, P62;SQSTM1, p62B, PDB3, sequestosome 1, SQSTM1, Ubiquitin binding protein p62, ZIP3
SQSTM1
IgG2a
Protein A
Antibody
1H5C1
Immunocytochemistry, Immunofluorescence, Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Human
Q13501
8878
P62/SQSTM1 Fusion Protein Ag13131
Primary
-20°C
SQSTM1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.