missing translation for 'onlineSavingsMsg'
Learn More

p62/SQSTM1 Antibody, DyLight 594, Novus Biologicals™

Product Code. 18416763 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416763 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416763 Supplier Novus Biologicals Supplier No. NBP149954DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

p62/SQSTM1 Polyclonal antibody specifically detects p62/SQSTM1 in Human, Mouse, Canine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen p62/SQSTM1
Applications Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias A170, Autophagy Receptor P62, DMRV, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, FTDALS3, NADGP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, PDB3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 325 and 375 of human p62/SQSTM1 using the numbering given in entry NP_003891.1 (GeneID 8878).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Chromatin Modifiers, Chromatin Research, DNA Methyltransferases, Epigenetics, Innate Immunity, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8878
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.