missing translation for 'onlineSavingsMsg'
Learn More

p62/SQSTM1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications

£188.00 - £365.00

Specifications

Antigen p62/SQSTM1
Concentration 1 mg/mL
Dilution Western Blot 1:2000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 35451674), Immunocytochemistry/Immunofluorescence 1:50, Knockout Validated
Applications Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18049275
View Documents
Novus Biologicals
NBP1-42822SS
0.025 mL
£188.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18789753
View Documents
Novus Biologicals
NBP1-42822
£365.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications

Specifications

p62/SQSTM1
Western Blot 1:2000, Simple Western 1:25, Flow Cytometry reported in scientific literature (PMID 35451674), Immunocytochemistry/Immunofluorescence 1:50, Knockout Validated
Polyclonal
Rabbit
Autophagy, Signal Transduction
PBS and 30% Glycerol with 0.1% Sodium Azide
A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
SQSTM1
IgG
Affinity Purified
1 mg/mL
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
Unconjugated
RUO
Human, Mouse, Rat
Q13501
8878
A synthetic peptide made to an internal region of the human p62/SQSTM1 protein (within residues 400-450). [Swiss-Prot Q13501]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.