missing translation for 'onlineSavingsMsg'
Learn More

p57Kip2 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. 16822295 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16822295 20 μL 20µL
16812295 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16822295 Supplier Proteintech Supplier No. 233171AP20UL
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.

Specifications

Antigen p57Kip2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CDKN1C
Gene Accession No. P49918
Gene Alias BWCR, BWS, CDKN1C, KIP2, p57, p57Kip2, WBS
Gene Symbols CDKN1C
Host Species Rabbit
Immunogen p57Kip2 Fusion Protein Ag19849
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1028
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.