missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p57 Kip2 Recombinant Rabbit Monoclonal Antibody (JE02-08)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556534
This item is not returnable.
View return policy
Description
p57 Kip2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| p57 Kip2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P49918, P49919 | |
| CDKN1C | |
| Synthetic peptide within human p57 kip2 aa 31-80/316. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE02-08 | |
| Unconjugated | |
| CDKN1C | |
| AL024410; Beckwith Wiedemann syndrome (WBS); BWCR; BWS; CDKI; Cdkn1c; cyclin dependent kinase inhibitor 1C; Cyclin dependent kinase inhibitor 1C (CDKN1C); cyclin-dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C (P57); cyclin-dependent kinase inhibitor 1C (p57, Kip2); cyclin-dependent kinase inhibitor p57; IMAGE; KIP2; p57; p57 Kip2; p57(kip2); p57KIP2; p57KIP2 A3sh; WBS | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1028, 12577, 246060 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction