missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p57 Kip2 Monoclonal Antibody (KP39), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA514361
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
MA5-14361 targets p57Kip2 in IHC (P) and IP applications and shows reactivity with Human and mouse samples. The MA5-14361 immunogen is recombinant human p57Kip2 protein.
This gene is part of a gene cluster on chromosome Xp11.23. The encoded protein contains a zinc finger motif often found in transcriptional regulators, however, its exact function is not known. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifica
| p57 Kip2 | |
| Monoclonal | |
| 0.05 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P49918, P49919 | |
| CDKN1C | |
| Recombinant human p57Kip2 protein | |
| 500 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| KP39 | |
| Unconjugated | |
| CDKN1C | |
| AL024410; Beckwith Wiedemann syndrome (WBS); BWCR; BWS; CDKI; Cdkn1c; cyclin dependent kinase inhibitor 1C; Cyclin dependent kinase inhibitor 1C (CDKN1C); cyclin-dependent kinase inhibitor 1C; cyclin-dependent kinase inhibitor 1C (P57); cyclin-dependent kinase inhibitor 1C (p57, Kip2); cyclin-dependent kinase inhibitor p57; IMAGE; KIP2; p57; p57 Kip2; p57(kip2); p57KIP2; p57KIP2 A3sh; WBS | |
| Mouse | |
| Protein A | |
| RUO | |
| 1028, 12577 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction