missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P54 of ASFV Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
£119.00 - £295.00
Specifications
| Antigen | P54 of ASFV |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235322
|
Proteintech
29511-1-AP-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233471
|
Proteintech
29511-1-AP-150UL |
150 μL |
£349.00 £295.00 / 150µL Save £54.00 15% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The P54 of ASFV antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of asfv P54 of ASFV. This antibody recognizes human antigen. The P54 of ASFV antibody has been validated for the following applications: ELISA analysis.Specifications
| P54 of ASFV | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1000 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 22220355 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title