missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p53R2 Recombinant Mouse Monoclonal Antibody (K1H015_6B3)
GREENER_CHOICE

Product Code. 30829338
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829338 100 μL
30826644 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829338 Supplier Invitrogen™ Supplier No. MA564463

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: GLDQDERSSS DTNESEIKSN EEPLLRKSSR RFVIFPIQYP DIWKMYKQAQ ASFWTAEEVD LSKDLPHWNK LKADEKYFIS HILAFFAASD GIVNENLVER FSQEVQVPEA RCFYGFQILI ENVHSEMYSL LIDTYIRDPK KREFLFNAIE TMPYVKKKAD WALRWIADRK Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 81%, Pig - 93%, Cynomolgus monkey - 98%.

The p53 tumor-suppressor gene integrates numerous signals that control cell life and death. Several novel molecules involved in p53 signaling, including p53R2, Chk2, p53AIP1, Noxa, PIDD, and PID/MTA2, were recently discovered. p53R2 is a p53 inducible gene that contains a p53 binding sequence and encodes a subunit of the enzyme ribonucleotide reductase. p53R2 is induced by the reagents, ultraviolet and -gamma-irradiation that cause DNA damages. The product of p53R2 gene is directly involved in the p53 checkpoint for repair of damaged DNA. The isoform of the p53 family member p73 also induces p53R2 expression. p53R2 is an important target of p53 for tumor suppression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53R2
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K1H015_6B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Rrm2b
Gene Accession No. Q7LG56
Gene Alias MTDPS8A; MTDPS8B; p53-inducible ribonucleotide reductase small subunit 2 homolog; p53-inducible ribonucleotide reductase small subunit 2 short form beta; p53-inducible ribonucleotide reductase small subunit 2-like protein; p53R2; Ribonucleoside-diphosphate reductase subunit M2 B; ribonucleotide reductase M2 B (TP53 inducible); ribonucleotide reductase regulatory TP53 inducible subunit M2B; RRM2B; TP53-inducible ribonucleotide reductase M2 B
Gene Symbols Rrm2b
Host Species Mouse
Immunogen Recombinant human RRM2B (aa 11-180).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50484
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.