missing translation for 'onlineSavingsMsg'
Learn More

p53R2 Antibody (001), DyLight 755, Novus Biologicals™

Product Code. 18376051 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376051 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376051 Supplier Novus Biologicals Supplier No. NBP290185IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

p53R2 Monoclonal specifically detects p53R2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53R2
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias EC 1.17.4.1, MGC102856, MGC42116, MTDPS8A, MTDPS8B, p53-inducible ribonucleotide reductase small subunit 2 homolog, p53-inducible ribonucleotide reductase small subunit 2-like protein, p53R2, P53R2DKFZp686M05248, PEOA5, ribonucleoside-diphosphate reductase subunit M2 B, ribonucleotide reductase M2 B (TP53 inducible), TP53-inducible ribonucleotide reductase M2 B
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human p53R2 (Q7LG56-1; Met1-Phe351).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Hypoxia, Modulation of DNA Pools
Primary or Secondary Primary
Gene ID (Entrez) 50484
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.