missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ p53DINP1 Recombinant Rabbit Monoclonal Antibody (K01_3K59)

Product Code. 30830030
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830030 100 μL
30825065 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830030 Supplier Invitrogen™ Supplier No. MA564364
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

p53DINP1 Recombinant Monoclonal Antibody for Western Blot

A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen p53DINP1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_3K59
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene TP53INP1
Gene Accession No. Q80YE2, Q96A56, Q9QXE4
Gene Alias 2700057G22Rik; p53-dependent damage-inducible nuclear protein 1; p53DINP1; p53-inducible p53DINP1; Sip; SIP18; SIP27; Stinp; stress induced protein; stress-induced protein; TEAP; thymus expressed acidic protein; thymus-expressed acidic protein; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; transformation related protein 53 inducible nuclear protein 1; Trp53inp1; tumor protein p53 inducible nuclear protein 1; tumor protein p53-inducible nuclear protein 1
Gene Symbols TP53INP1, Trp53inp1
Host Species Rabbit
Immunogen Peptide.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297822, 60599, 94241
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.