missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p53DINP1 Recombinant Rabbit Monoclonal Antibody (9Y6Z7)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542878
This item is not returnable.
View return policy
Description
Positive test controls include: HepG2, A-549, BxPC-3, RAW264.7, C6, Mouse lung, Mouse liver, Rat lung. Immunogen sequence: PPPCFTAGGL TTIKVETSPM ENLLIEHPSM SVYAVHNSCP GLSEATRGTD ELHSPSSPRV EAQNEMGQHI HCYVAALAAH TTFLEQPKSF RPSQWIKEHS E.
A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
Specifications
| p53DINP1 | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80YE2, Q96A56, Q9QXE4 | |
| TP53INP1, Trp53inp1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human p53 DINP1 (Q96A56). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 9Y6Z7 | |
| Unconjugated | |
| TP53INP1 | |
| 2700057G22Rik; p53-dependent damage-inducible nuclear protein 1; p53DINP1; p53-inducible p53DINP1; Sip; SIP18; SIP27; Stinp; stress induced protein; stress-induced protein; TEAP; thymus expressed acidic protein; thymus-expressed acidic protein; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; transformation related protein 53 inducible nuclear protein 1; Trp53inp1; tumor protein p53 inducible nuclear protein 1; tumor protein p53-inducible nuclear protein 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 297822, 60599, 94241 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction