missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p53DINP1 Polyclonal Antibody

Description
In Western blot applications, this antibody detects a band at ~27kDa. A suggested positive control is human lung tissue lysate. PA5-20009 can be used with blocking peptide PEP-0129.
A novel p53 inducible gene was identified and designated p53DINP1 (for p53-dependent damage-inducible nuclear protein). p53DINP1 may regulate p53-dependent apoptosis through phosphorylation at Ser46 and induction of p53AIP1. p53DINP1/SIP is expressed in many tissues and induced by a variety of stress agents including UV stress, mutagenic stress, heat shock, and oxidative stress.
Specifications
Specifications
| Antigen | p53DINP1 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | TP53INP1 |
| Gene Accession No. | Q80YE2, Q96A56, Q9QXE4 |
| Gene Alias | 2700057G22Rik; p53-dependent damage-inducible nuclear protein 1; p53DINP1; p53-inducible p53DINP1; Sip; SIP18; SIP27; Stinp; stress induced protein; stress-induced protein; TEAP; thymus expressed acidic protein; thymus-expressed acidic protein; TP53DINP1; TP53INP1; TP53INP1A; TP53INP1B; transformation related protein 53 inducible nuclear protein 1; Trp53inp1; tumor protein p53 inducible nuclear protein 1; tumor protein p53-inducible nuclear protein 1 |
| Gene Symbols | TP53INP1, Trp53inp1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?