missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p53 Tumor Suppressor Protein Mouse, Clone: PCRP-TP53-2A10, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7157-MSM14-P1
This item is not returnable.
View return policy
Specifications
| p53 Tumor Suppressor Protein | |
| Nuclear | |
| PCRP-TP53-2A10 | |
| Unconjugated | |
| Mouse | |
| 53kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 7157 | |
| IgG2b |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human TP53 protein | |
| HeLa cells. Colorectal or Ovarian Carcinoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction