missing translation for 'onlineSavingsMsg'
Learn More
Learn More
P53 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
£119.00 - £330.00
Specifications
| Antigen | P53 |
|---|---|
| Clone | 2H13 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234580
|
Proteintech
80077-1-RR-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234334
|
Proteintech
80077-1-RR-100UL |
100 μL |
£360.00 £330.00 / 100µL Save £30.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The P53 antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human P53. This antibody recognizes human, mouse, rat, zebrafish antigen. The P53 antibody has been validated for the following applications: WB, IHC, IF/ICC, FC (Intra), ELISA analysis.Specifications
| P53 | |
| 500 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 7157 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 2H13 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Zebrafish | |
| FLJ92943; LFS1; TRP53; p53 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title