Learn More
P53 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL594-10442-100UL
Description
The P53 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human P53. This antibody recognizes human, rat, mouse antigen. The P53 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| P53 | |
| Polyclonal | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |
| Immunofluorescence | |
| CoraLite 594 | |
| 100 μL | |
| 7157 | |
| IgG |