missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ p53 (R175H Mutant) Recombinant Rabbit Monoclonal Antibody (HL1129)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546935
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
This gene encodes a tumor suppressor protein containing transcriptional activation, DNA binding, and oligomerization domains The encoded protein responds to diverse cellular stresses to regulate expression of target genes, thereby inducing cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism Mutations in this gene are associated with a variety of human cancers, including hereditary cancers such as Li-Fraumeni syndrome Alternative splicing of this gene and the use of alternate promoters result in multiple transcript variants and isoforms Additional isoforms have also been shown to result from the use of alternate translation initiation codons from identical transcript variants (PMIDs: 12032546, 20937277).
Specifications
| p53 (R175H Mutant) | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| BCC7; BMFS5; LFS1; P53; TRP53; tumor protein p53 | |
| Carrier-protein conjugated synthetic peptide surrounding mutant R175H of human p53. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot | |
| HL1129 | |
| Unconjugated | |
| P29466, P48729, Q8N752 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 122011, 1452, 834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction