Learn More
p53, p Ser15 Antibody (261366), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB18391E
Product Code. 18566908
  • £516.00 / 0.10mL
Description
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
p53
Monoclonal
DyLight 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
261366
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53
Phosphopeptide containing the human p53 S15 site, Accession # P04637
0.1 mL
Apoptosis, Cancer, Cell Cycle and Replication, Cellular Markers, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, p53 Pathway, Phospho Specific, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors
7157
Store at 4°C in the dark.
IgG1