missing translation for 'onlineSavingsMsg'
Learn More

p53 and MDM2 proteins-interaction-inhibitor dihydrochloride, MedChemExpress

Product Code. 16467926
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467926 100 mg 100mg
16408178 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16467926 Supplier Medchem Express Supplier No. HY70027A
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress p53 and MDM2 proteins-interaction-inhibitor dihydrochloride is an inhibitor of the interaction between p53 and MDM2 proteins.

p53 and MDM2 proteins-interaction-inhibitor dihydrochloride is an inhibitor of the interaction between p53 and MDM2 proteins.

Specifications

Chemical Name or Material p53 and MDM2 proteins-interaction-inhibitor dihydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C40H51Cl4N5O4
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (123.81 mM; Need ultrasonic)
SMILES O=C(N1CCN(CC(N2CCOCC2)=O)CC1)N3[C@](C)(C4=CC=C(Cl)C=C4)[C@](C)(C5=CC=C(Cl)C=C5)N=C3C6=CC=C(C(C)(C)C)C=C6OCC.Cl.Cl
Molecular Weight (g/mol) 807.68
Formula Weight 807.68
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.